Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERTAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013368.3 | NP_037500.2 | 2 | 549 | ||
NM_203344.2 | NP_976219.1 | 2 | 438 | |||
XM_005258832.3 | XP_005258889.1 | 3 | 646 | |||
XM_006723179.3 | XP_006723242.1 | 2 | 498 | |||
GenBank mRNA | AF192529.1 | - | 2 | 562 | ||
AK074604.1 | - | 2 | 363 | |||
BC014061.2 | - | 2 | 362 | 3926937 | ||
BC050643.2 | - | 2 | 438 | 6503515 |