Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: DNMT3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013369.3 | NP_037501.2 | 10 | 1327 | ||
NM_175867.2 | NP_787063.1 | 10 | 1327 | |||
GenBank mRNA | AF194032.1 | - | 10 | 1327 | ||
BC002560.2 | - | 10 | 1001 | 3138514 | ||
BQ028660.1 | - | 3 | 390 |
Gene Symbol: LOC105372833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135514.1 | 1 | 60 | |||
GenBank mRNA | AI631758.1 | - | 2 | 468 |