Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STMN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276310.1 | NP_001263239.1 | 2, 3 | 301 | ||
NM_015894.3 | NP_056978.2 | 2, 3 | 391 | |||
NR_075070.1 | 2, 3 | 391 | ||||
GenBank mRNA | AK074860.1 | - | 2, 3 | 203 | ||
AK094112.1 | - | 2, 3 | 826 | |||
AK298903.1 | - | 2, 3 | 301 | |||
AK310010.1 | - | 2, 3 | 226 | |||
BC009381.2 | - | 2, 3 | 248 | 4124704 | ||
BC025234.1 | - | 2, 3 | 248 | 4894236 | ||
BC047671.1 | - | 2, 3 | 138 | |||
DA161621.1 | - | 2, 3 | 391 | |||
DA181040.1 | - | 2, 3 | 375 |