Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF20L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277196.1 | NP_001264125.1 | 6 | 947 | ||
NM_016018.4 | NP_057102.4 | 7 | 1025 | |||
NM_198513.1 | NP_940915.1 | 6 | 943 | |||
XM_005250931.1 | XP_005250988.1 | 7 | 1007 | |||
XM_011517075.1 | XP_011515377.1 | 7 | 1007 | |||
XM_011517076.1 | XP_011515378.1 | 7 | 1003 | |||
XM_011517077.1 | XP_011515379.1 | 7 | 1007 | |||
XM_011517078.1 | XP_011515380.1 | 7 | 995 | |||
XM_011517079.1 | XP_011515381.1 | 7 | 995 | |||
XM_011517080.1 | XP_011515382.1 | 6 | 917 | |||
XM_011517081.1 | XP_011515383.1 | 6 | 917 | |||
XM_011517088.1 | XP_011515390.1 | 7 | 1007 | |||
XM_017013509.1 | XP_016868998.1 | 7 | 991 | |||
XM_017013510.1 | XP_016868999.1 | 6 | 913 | |||
XM_017013511.1 | XP_016869000.1 | 6 | 913 | |||
XM_017013512.1 | XP_016869001.1 | 6 | 913 | |||
XM_017013513.1 | XP_016869002.1 | 7 | 3830 | |||
XM_017013514.1 | XP_016869003.1 | 7 | 3826 | |||
XM_017013516.1 | XP_016869005.1 | 7 | 995 | |||
XM_017013517.1 | XP_016869006.1 | 6 | 917 | |||
GenBank mRNA | AB101203.1 | - | 7 | 992 | ||
AF151830.1 | - | 4 | 379 | |||
AK022280.1 | - | 5 | 628 | |||
AK023711.1 | - | 1 | 470 | |||
AK290506.1 | - | 6 | 917 | |||
BC002822.2 | - | 7 | 983 | |||
BC015211.1 | - | 6 | 893 |