Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016034.4 | NP_057118.1 | 4 | 712 | ||
NR_051967.1 | 5 | 855 | ||||
NR_051968.1 | 5 | 910 | ||||
NR_051969.1 | 4 | 915 | ||||
NR_051970.1 | 4 | 857 | ||||
XM_006717136.3 | XP_006717199.1 | 5 | 836 | |||
GenBank mRNA | AB055388.1 | - | 4 | 636 | ||
AF151849.1 | - | 4 | 636 | |||
BC004905.2 | - | 1 | 207 | |||
BC008017.1 | - | 4 | 648 | 3534388 | ||
BC013108.2 | - | 4 | 656 | 3924671 | ||
BX362434.2 | - | 4 | 706 |