Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016246.2 | NP_057330.2 | 7 | 780 | ||
XM_005258969.3 | XP_005259026.1 | 7 | 789 | |||
XM_005258970.3 | XP_005259027.1 | 6 | 699 | |||
GenBank mRNA | AF126781.1 | - | 7 | 626 | ||
AK307437.1 | - | 7 | 589 | |||
AY358430.1 | - | 7 | 619 | |||
BC006283.2 | - | 7 | 573 | 4026397 | ||
BC006294.1 | - | 7 | 581 | 4025809 |