Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320480.1 | NP_001307409.1 | 3 | 444 | ||
NM_016430.3 | NP_057514.2 | 4 | 458 | |||
NM_153681.2 | NP_710148.1 | 2 | 399 | |||
NM_153682.2 | NP_710149.1 | 3 | 292 | |||
XM_005260990.4 | XP_005261047.1 | 3 | 157 | |||
XM_011529596.2 | XP_011527898.1 | 3 | 191 | |||
XM_017028364.1 | XP_016883853.1 | 3 | 209 | |||
XM_017028365.1 | XP_016883854.1 | 4 | 400 | |||
GenBank mRNA | AB035742.1 | - | 3 | 224 | ||
AB035743.1 | - | 3 | 154 | |||
AB035744.1 | - | 3 | 186 | |||
AB035745.1 | - | 2 | 206 | |||
AB037162.1 | - | 3 | 228 | |||
AB037163.1 | - | 2 | 197 | |||
AB037164.1 | - | 3 | 309 | |||
AB039659.1 | - | 2 | 111 | |||
AB451328.1 | - | 1 | 33 | |||
AB451472.1 | - | 1 | 33 | |||
AF216305.1 | - | 4 | 374 | |||
AF237812.1 | - | 2 | 248 | |||
AK026311.1 | - | 2 | 184 | |||
AK304656.1 | - | 2 | 318 | |||
AK314457.1 | - | 2 | 192 | |||
AK316609.1 | - | 2 | 129 | |||
BC005180.1 | - | 3 | 173 | 3929930 | ||
BC011007.1 | - | 3 | 177 | 4281552 | ||
BE275469.1 | - | 3 | 291 | |||
BT007053.1 | - | 2 | 111 |