Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165895.1 | NP_001159367.1 | 2, 3 | 439 | ||
NM_016579.3 | NP_057663.1 | 3, 4 | 565 | |||
GenBank mRNA | AF161254.1 | - | 3, 4 | 564 | ||
AK058014.1 | - | 2, 3 | 433 | |||
AK222623.1 | - | 3, 4 | 549 | |||
AK315656.1 | - | 3, 4 | 550 | |||
AL136652.1 | - | 3, 4 | 554 | |||
AL365455.1 | - | 3, 4 | 537 | |||
AY358420.1 | - | 3, 4 | 531 | |||
BC000668.2 | - | 3, 4 | 536 | 3347569 | ||
BC007083.1 | - | 3, 4 | 519 | 4076237 | ||
CR457174.1 | - | 3, 4 | 499 |