Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FKBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143781.1 | NP_001137253.1 | 4, 5 | 307 | ||
NM_016594.2 | NP_057678.1 | 5, 6 | 513 | |||
GenBank mRNA | AF238079.1 | - | 5, 6 | 466 | ||
AY358998.1 | - | 5, 6 | 471 | |||
BC027973.1 | - | 5, 6 | 465 | 5229643 | ||
BE857354.1 | - | 1, 2 | 224 |