Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRAID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170795.1 | NP_001164266.1 | 7 | 641 | ||
NM_016085.4 | NP_057169.2 | 7 | 781 | |||
NM_080592.3 | NP_542159.3 | 7 | 961 | |||
GenBank mRNA | AA604432.1 | - | 1 | 272 | ||
AF077037.1 | - | 7 | 643 | |||
AF144055.2 | - | 7 | 772 | |||
AF275744.1 | - | 8 | 635 | |||
AK291894.1 | - | 7 | 771 | |||
AW966996.1 | - | 166 | ||||
AY358968.1 | - | 7 | 634 | |||
BC002846.1 | - | 7 | 629 | |||
BC011006.1 | - | 7 | 635 | |||
BC021237.2 | - | 7 | 741 | 4303513 | ||
BC035850.1 | - | 7 | 627 | 5753421 | ||
BQ420456.1 | - | 7 | 764 | |||
BQ718371.1 | - | 7 | 676 | |||
CA450289.1 | - | 1 | 290 | |||
CD513162.1 | - | 2 | 138 |