Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015964.3 | NP_057048.2 | 4 | 570 | ||
NM_016140.3 | NP_057224.2 | 5 | 661 | |||
GenBank mRNA | AF078846.1 | - | 4 | 501 | ||
AF132972.1 | - | 4 | 490 | |||
BC000691.2 | - | 4 | 517 | 3349849 | ||
BI754230.1 | - | 5 | 644 |