Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282225.1 | NP_001269154.1 | 5 | 1123 | ||
NM_001282226.1 | NP_001269155.1 | 5 | 1090 | |||
NM_001282227.1 | NP_001269156.1 | 5 | 896 | |||
NM_001282228.1 | NP_001269157.1 | 5 | 998 | |||
NM_001282229.1 | NP_001269158.1 | 4 | 755 | |||
NM_177405.2 | NP_803124.1 | 2 | 329 | |||
XM_006724080.3 | XP_006724143.1 | 5 | 710 | |||
XM_011546133.1 | XP_011544435.1 | 5 | 1015 | |||
GenBank mRNA | AB209648.1 | - | 5 | 710 | ||
AF190746.1 | - | 4 | 1059 | |||
AK027682.1 | - | 2 | 150 | |||
AK074702.1 | - | 2 | 201 | |||
AK292689.1 | - | 5 | 1111 | |||
AK295174.1 | - | 5 | 847 | |||
AK304818.1 | - | 5 | 890 | |||
AK314321.1 | - | 5 | 914 | |||
BC051755.1 | - | 4 | 894 | 5753141 | ||
CR456417.1 | - | 4 | 881 | |||
DA864284.1 | - | 2 | 196 | |||
DB130953.1 | - | 2 | 329 |