Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282225.1 | NP_001269154.1 | 6 | 1181 | ||
NM_001282226.1 | NP_001269155.1 | 6 | 1148 | |||
NM_001282227.1 | NP_001269156.1 | 6 | 954 | |||
NM_001282228.1 | NP_001269157.1 | 6 | 1056 | |||
NM_001282229.1 | NP_001269158.1 | 5 | 813 | |||
NM_177405.2 | NP_803124.1 | 3 | 387 | |||
XM_006724080.3 | XP_006724143.1 | 5 | 2418 | |||
XM_011546133.1 | XP_011544435.1 | 6 | 1073 | |||
GenBank mRNA | AB209648.1 | - | 5 | 2418 | ||
AF190746.1 | - | 5 | 1117 | |||
AK027682.1 | - | 3 | 208 | |||
AK074702.1 | - | 3 | 259 | |||
AK292689.1 | - | 6 | 1169 | |||
AK304818.1 | - | 6 | 948 | |||
AK314321.1 | - | 6 | 972 | |||
BC051755.1 | - | 5 | 952 | 5753141 | ||
CR456417.1 | - | 5 | 939 | |||
DA864284.1 | - | 3 | 254 | |||
DB130953.1 | - | 3 | 387 |