Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001895.2 | NP_001001895.1 | 12, 13 | 1709 | ||
NM_018961.3 | NP_061834.1 | 13, 14 | 1823 | |||
XM_006724013.3 | XP_006724076.1 | 13, 14 | 1907 | |||
XM_011529606.2 | XP_011527908.1 | 13, 14 | 1792 | |||
XM_011529607.2 | XP_011527909.1 | 13, 14 | 1876 | |||
XM_011529610.2 | XP_011527912.1 | 9, 10 | 3822 | |||
GenBank mRNA | AF521702.1 | - | 12, 13 | 1636 | ||
AJ277750.1 | - | 13, 14 | 1770 | |||
BC028138.1 | - | 12, 13 | 2177 | 5215324 | ||
BC069357.1 | - | 13, 14 | 1770 | 7262315 | ||
BC069483.1 | - | 13, 14 | 1770 | 7262391 |