Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBASH3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001895.2 | NP_001001895.1 | 14 | 1881 | ||
NM_001243467.1 | NP_001230396.1 | 12 | 1664 | |||
NM_018961.3 | NP_061834.1 | 15 | 1995 | |||
XM_006724013.3 | XP_006724076.1 | 15 | 2079 | |||
XM_011529606.2 | XP_011527908.1 | 15 | 1964 | |||
XM_011529607.2 | XP_011527909.1 | 15 | 2048 | |||
XM_011529610.2 | XP_011527912.1 | 11 | 3994 | |||
GenBank mRNA | AF520809.1 | - | 15 | 1922 | ||
AF521702.1 | - | 14 | 1808 | |||
AJ277750.1 | - | 15 | 1942 | |||
AK308151.1 | - | 14 | 1822 | |||
AK313367.1 | - | 14 | 2020 | |||
BC028138.1 | - | 14 | 2349 | 5215324 | ||
BC069483.1 | - | 15 | 1942 | 7262391 | ||
BC069511.1 | - | 14 | 1826 | 7262403 | ||
BC069577.1 | - | 12 | 1611 |