Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM134B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034850.2 | NP_001030022.1 | 9 | 1252 | ||
NM_019000.4 | NP_061873.2 | 7 | 1128 | |||
XM_011514053.2 | XP_011512355.1 | 10 | 1311 | |||
XM_011514054.2 | XP_011512356.1 | 8 | 1262 | |||
XM_011514055.2 | XP_011512357.1 | 7 | 1052 | |||
GenBank mRNA | AK000159.1 | - | 1 | 171 | ||
AK024920.1 | - | 7 | 958 | |||
AK025808.1 | - | 9 | 1179 | |||
AK025832.1 | - | 1 | 174 | |||
AK225371.1 | - | 1 | 174 | |||
AL832438.1 | - | 9 | 928 | |||
BC030517.1 | - | 3 | 324 | |||
BC053326.1 | - | 9 | 1176 | 5014868 | ||
BC073132.1 | - | 7 | 961 | 5760145 | ||
CR994722.1 | - | 3 | 367 |