Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256164.1 | NP_001243093.1 | 7 | 620 | ||
NM_001256165.1 | NP_001243094.1 | 7 | 922 | |||
NM_019026.4 | NP_061899.2 | 7 | 722 | |||
NR_045818.1 | 7 | 645 | ||||
GenBank mRNA | AB020980.1 | - | 7 | 689 | ||
AF070626.1 | - | 7 | 678 | |||
AF274935.1 | - | 6 | 497 | |||
AK298614.1 | - | 7 | 889 | |||
AK316080.1 | - | 7 | 855 | |||
AK316610.1 | - | 7 | 668 | |||
AY359027.1 | - | 7 | 657 | |||
AY763589.1 | - | 7 | 651 | |||
BC000104.2 | - | 7 | 642 | 3504808 |