Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EQTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161585.1 | NP_001155057.1 | 6 | 576 | ||
NM_020641.2 | NP_065692.2 | 7 | 663 | |||
XM_011517920.2 | XP_011516222.1 | 4 | 1342 | |||
GenBank mRNA | AJ278482.1 | - | 7 | 663 | ||
BC137351.1 | - | 7 | 633 | 9021349 | ||
BC137352.1 | - | 7 | 633 | 9021350 | ||
BC144602.1 | - | 6 | 546 | 9053133 | ||
BX089774.1 | - | 1 | 48 |