Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APTX SIRNA Assays ›
Gene Symbol:
APTX
Gene Name:
aprataxin
Gene Aliases:
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
Chromosome Location:
Chr.9: 32883872 - 33025129 on Build GRCh38
UniGene ID:
Hs.20158
Species:
Human
siRNA ID
HSS147791
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APTX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195248.1
NP_001182177.1
3
240
NM_001195249.1
NP_001182178.1
3
409
NM_001195251.1
NP_001182180.1
4
346
NM_001195252.1
NP_001182181.1
3
240
NM_175069.2
NP_778239.1
3
240
NM_175073.2
NP_778243.1
4
346
NR_036576.1
3
235
NR_036577.1
3
240
NR_036578.1
3
257
NR_036579.1
3
404
XM_006716791.3
XP_006716854.1
3
248
XM_006716792.2
XP_006716855.1
3
266
XM_017014831.1
XP_016870320.1
3
240
XM_017014832.1
XP_016870321.1
3
240
XM_017014833.1
XP_016870322.1
3
240
XM_017014834.1
XP_016870323.1
3
240
XM_017014835.1
XP_016870324.1
3
240
XM_017014836.1
XP_016870325.1
3
240
XM_017014838.1
XP_016870327.1
3
222
GenBank mRNA
AB603741.1
-
2
136
AJ565850.1
-
3
213
AJ565851.1
-
3
218
AJ565852.1
-
3
213
AJ565853.1
-
2
216
AJ565854.1
-
2
221
AJ575566.1
-
2
216
AK000164.1
-
2
358
AK055672.1
-
3
227
AK131046.1
-
3
192
AY040777.1
-
3
215
AY208829.1
-
3
215
AY208831.1
-
3
206
AY208834.1
-
3
211
AY208835.1
-
3
195
AY208836.1
-
3
355
AY208837.1
-
4
312
AY208838.1
-
2
286
AY208839.1
-
3
349
AY208840.1
-
3
212
AY208841.1
-
2
164
AY302067.1
-
2
358
AY302068.1
-
2
353
AY302069.1
-
2
358
AY302070.1
-
2
353
AY302071.1
-
2
358
AY302072.1
-
2
353
AY302073.1
-
2
358
AY302074.1
-
2
353
BC001628.1
-
2
178
BC032650.1
-
3
281
5505120
BC068107.1
-
4
321
BC104881.1
-
3
254
8143884
BQ957536.1
-
4
329
BX538161.1
-
4
325
DA664623.1
-
2
267
Back To Top
More Information
Panther Classification:
Molecular Function -
damaged DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
DNA ligation
cellular response to DNA damage stimulus
dephosphorylation
regulation of protein stability
response to hydrogen peroxide
nucleic acid phosphodiester bond hydrolysis
chromatin binding
damaged DNA binding
double-stranded DNA binding
double-stranded RNA binding
protein binding
phosphoglycolate phosphatase activity
DNA 5'-adenosine monophosphate hydrolase activity
metal ion binding
protein N-terminus binding
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0