Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5SL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167867.1 | NP_001161339.1 | 6 | 775 | ||
NM_001167868.1 | NP_001161340.1 | 5 | 612 | |||
NM_001167869.1 | NP_001161341.1 | 5 | 566 | |||
NM_001167870.1 | NP_001161342.1 | 4 | 485 | |||
NM_001167871.1 | NP_001161343.1 | 5 | 648 | |||
NM_001320838.1 | NP_001307767.1 | 5 | 529 | |||
NM_001320840.1 | NP_001307769.1 | 5 | 694 | |||
NM_001320841.1 | NP_001307770.1 | 4 | 531 | |||
NM_018035.2 | NP_060505.2 | 6 | 729 | |||
NR_030765.1 | 6 | 712 | ||||
NR_135476.1 | 5 | 580 | ||||
NR_135477.1 | 6 | 726 | ||||
NR_135478.1 | 6 | 743 | ||||
XM_011527065.2 | XP_011525367.1 | 6 | 780 | |||
GenBank mRNA | AK001103.1 | - | 6 | 696 | ||
AK123631.1 | - | 3 | 1335 | |||
AK293132.1 | - | 5 | 575 | |||
AK297698.1 | - | 5 | 536 | |||
AK298197.1 | - | 5 | 647 | |||
AK301940.1 | - | 6 | 738 | |||
AK316313.1 | - | 5 | 621 | |||
BC013323.1 | - | 4 | 461 | 4133534 | ||
BQ072604.1 | - | 6 | 701 | |||
DA540661.1 | - | 5 | 497 | |||
DA631852.1 | - | 5 | 550 |