Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300926.1 | NP_001287855.1 | 2 | 437 | ||
NM_018320.4 | NP_060790.2 | 3 | 475 | |||
NR_024147.1 | 2 | 437 | ||||
NR_024148.1 | 2 | 437 | ||||
XM_006718629.1 | XP_006718692.1 | 3 | 496 | |||
XM_006718630.1 | XP_006718693.1 | 3 | 150 | |||
XM_006718632.1 | XP_006718695.1 | 3 | 496 | |||
XM_011545150.1 | XP_011543452.1 | 3 | 170 | |||
GenBank mRNA | AK023139.1 | - | 3 | 141 | ||
AK094508.1 | - | 2 | 103 | |||
AL833638.1 | - | 2 | 112 | |||
BC009672.2 | - | 3 | 143 | |||
BC093084.1 | - | 2 | 77 | |||
BC117273.1 | - | 3 | 107 | |||
BG428563.1 | - | 3 | 163 | |||
BX463021.2 | - | 2 | 126 | |||
CR457298.1 | - | 3 | 135 | |||
DA209781.1 | - | 3 | 496 | |||
DA694800.1 | - | 3 | 151 |