Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300926.1 | NP_001287855.1 | 7 | 1077 | ||
NM_018320.4 | NP_060790.2 | 9 | 1239 | |||
NR_024147.1 | 8 | 1201 | ||||
NR_024148.1 | 9 | 1339 | ||||
XM_006718629.1 | XP_006718692.1 | 8 | 1152 | |||
XM_006718630.1 | XP_006718693.1 | 10 | 1052 | |||
XM_006718632.1 | XP_006718695.1 | 7 | 997 | |||
XM_011545150.1 | XP_011543452.1 | 10 | 1084 | |||
GenBank mRNA | AK001961.1 | - | 4 | 403 | ||
AK023139.1 | - | 9 | 905 | |||
AK094508.1 | - | 9 | 1005 | |||
AL833638.1 | - | 7 | 752 | |||
BC009672.2 | - | 9 | 907 | |||
BC063680.1 | - | 4 | 415 | |||
BC093084.1 | - | 8 | 841 | |||
BC117273.1 | - | 9 | 871 | |||
BG328761.1 | - | 3 | 211 | |||
BX463021.2 | - | 5 | 506 | |||
CR457298.1 | - | 9 | 899 |