Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291702.1 | NP_001278631.1 | 2 | 387 | ||
NM_001291703.1 | NP_001278632.1 | 2 | 387 | |||
GenBank mRNA | AB435052.1 | - | 2 | 314 | ||
AB435053.1 | - | 2 | 314 | |||
AB435054.1 | - | 2 | 314 | |||
AB435055.1 | - | 2 | 314 | |||
AB435056.1 | - | 2 | 314 | |||
AB435057.1 | - | 2 | 314 | |||
AB435058.1 | - | 2 | 314 | |||
AB435059.1 | - | 2 | 314 | |||
AB435060.1 | - | 1 | 410 | |||
AB435061.1 | - | 2 | 314 | |||
AJ238982.1 | - | 2 | 359 | |||
AK075110.1 | - | 2 | 386 | |||
AK314708.1 | - | 2 | 387 | |||
BC104416.2 | - | 2 | 374 | |||
BC104417.1 | - | 2 | 374 | |||
BC113022.1 | - | 2 | 374 | |||
BC113023.1 | - | 2 | 374 | |||
BC148317.2 | - | 2 | 374 | 40015223 | ||
DA670661.1 | - | 2 | 388 |