Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018653.3 | NP_061123.3 | 2 | 1064 | ||
NM_022036.2 | NP_071319.2 | 2 | 1617 | |||
XM_005257512.1 | XP_005257569.1 | 2 | 1411 | |||
XM_005257514.1 | XP_005257571.1 | 2 | 1804 | |||
XM_017024855.1 | XP_016880344.1 | 2 | 1411 | |||
XM_017024856.1 | XP_016880345.1 | 2 | 1411 | |||
GenBank mRNA | AB451470.1 | - | 1 | 1007 | ||
AF207989.1 | - | 2 | 1053 | |||
AJ276102.1 | - | 2 | 1073 | |||
AK000249.1 | - | 3 | 163 | |||
AK075397.1 | - | 2 | 1055 | |||
AK131210.1 | - | 2 | 2232 | |||
AK222909.1 | - | 2 | 1035 | |||
BC016860.1 | - | 1 | 994 | |||
BC110848.1 | - | 2 | 1748 | 5588871 | ||
BC125080.1 | - | 2 | 1609 | 40117722 | ||
BC125081.1 | - | 2 | 1609 | 40117723 | ||
BT007435.1 | - | 1 | 971 |
Set Membership: |
![]() |