Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF302 SIRNA Assays ›
Gene Symbol:
ZNF302
Gene Name:
zinc finger protein 302
Gene Aliases:
HSD16, MST154, MSTP154, ZNF135L, ZNF140L, ZNF327
Chromosome Location:
Chr.19: 34677619 - 34686397 on Build GRCh38
UniGene ID:
Hs.436350
Species:
Human
siRNA ID
HSS148178
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF302
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001012320.2
NP_001012320.1
5
1868
NM_001289181.1
NP_001276110.1
6
2023
NM_001289182.1
NP_001276111.1
5
2141
NM_001289183.1
NP_001276112.1
5
1831
NM_001289184.1
NP_001276113.1
5
1894
NM_001289185.1
NP_001276114.1
5
1871
NM_001289186.1
NP_001276115.1
5
1789
NM_001289187.1
NP_001276116.1
5
2138
NM_001289188.1
NP_001276117.1
5
2160
NM_001289189.1
NP_001276118.1
5
2154
NM_001289190.1
NP_001276119.1
5
2091
NM_001289191.1
NP_001276120.1
5
2128
NM_001289192.1
NP_001276121.1
5
2151
NM_018443.3
NP_060913.2
5
1891
NM_018675.2
NP_061145.2
5
2398
NR_110322.1
3
1686
XM_011527111.2
XP_011525413.1
5
2151
XM_017026978.1
XP_016882467.1
6
2264
XM_017026979.1
XP_016882468.1
6
2261
XM_017026980.1
XP_016882469.1
6
1955
XM_017026981.1
XP_016882470.1
6
1918
XM_017026982.1
XP_016882471.1
5
1766
XM_017026986.1
XP_016882475.1
5
2392
GenBank mRNA
AF155656.1
-
5
1842
AF190157.1
-
5
1830
AF265236.1
-
6
2178
AF326206.1
-
5
1941
AK074901.1
-
5
1841
AK122855.1
-
3
3090
AL162002.1
-
1
231
AL833690.1
-
1
1175
AL834318.1
-
5
1845
AY500357.1
-
1
1126
BC024176.2
-
5
1943
4822400
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0