Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PCDHB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018930.3 | NP_061753.1 | 1 | 1216 | ||
GenBank mRNA | AF131761.1 | - | 1 | 39 | ||
AF152489.1 | - | 1 | 1042 | |||
AF217748.1 | - | 1 | 1042 | |||
AK056391.1 | - | 1 | 1226 | |||
AK315583.1 | - | 1 | 1242 | |||
AY358720.1 | - | 1 | 1220 | |||
BC031837.1 | - | 1 | 1216 | 4931995 |