Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021800.2 | NP_068572.1 | 3 | 378 | ||
NM_201262.1 | NP_957714.1 | 3 | 378 | |||
XM_017016431.1 | XP_016871920.1 | 2 | 150 | |||
XM_017016432.1 | XP_016871921.1 | 2 | 121 | |||
GenBank mRNA | AF176012.1 | - | 3 | 363 | ||
AF176013.1 | - | 3 | 295 | |||
AK289537.1 | - | 3 | 328 | |||
BC017018.1 | - | 3 | 314 | 3884093 | ||
CB121724.1 | - | 3 | 380 | |||
CR457184.1 | - | 3 | 210 |