Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037553.1 | NP_001032642.1 | 7 | 916 | ||
NM_020132.4 | NP_064517.1 | 8 | 1036 | |||
XM_005261160.4 | XP_005261217.1 | 8 | 1037 | |||
XM_006724029.3 | XP_006724092.1 | 7 | 965 | |||
XM_006724030.3 | XP_006724093.1 | 7 | 933 | |||
XM_006724031.3 | XP_006724094.1 | 7 | 936 | |||
XM_011529660.2 | XP_011527962.1 | 7 | 775 | |||
XM_011529661.2 | XP_011527963.1 | 7 | 1090 | |||
XM_011529662.2 | XP_011527964.1 | 9 | 1740 | |||
XM_011529663.2 | XP_011527965.1 | 7 | 949 | |||
XM_011529664.2 | XP_011527966.1 | 7 | 890 | |||
XM_011529665.2 | XP_011527967.1 | 9 | 1391 | |||
XM_017028408.1 | XP_016883897.1 | 7 | 919 | |||
XM_017028409.1 | XP_016883898.1 | 7 | 997 | |||
XM_017028410.1 | XP_016883899.1 | 7 | 1002 | |||
XM_017028411.1 | XP_016883900.1 | 7 | 875 | |||
XM_017028412.1 | XP_016883901.1 | 8 | 1968 | |||
XM_017028413.1 | XP_016883902.1 | 8 | 936 | |||
GenBank mRNA | AB040138.1 | - | 7 | 991 | ||
AF156774.1 | - | 8 | 997 | |||
AF156775.1 | - | 7 | 860 | |||
AK125804.1 | - | 5 | 3653 | |||
AL832919.1 | - | 5 | 608 | |||
AY358704.1 | - | 1085 | ||||
BC011971.1 | - | 8 | 1010 | 4148915 | ||
BC040603.1 | - | 9 | 1740 | 5270905 | ||
BC063552.1 | - | 7 | 1306 | 4800514 |