Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128626.1 | NP_001122098.1 | 17 | 2208 | ||
NM_001128627.1 | NP_001122099.1 | 16 | 1864 | |||
NM_020148.2 | NP_064533.3 | 16 | 2166 | |||
XM_005258122.4 | XP_005258179.1 | 17 | 2523 | |||
XM_011525701.1 | XP_011524003.1 | 18 | 2571 | |||
XM_011525702.1 | XP_011524004.1 | 18 | 2083 | |||
GenBank mRNA | AB032961.2 | - | 17 | 2260 | ||
AJ277587.1 | - | 14 | 1753 | |||
AK025433.1 | - | 1 | 122 | |||
AK290180.1 | - | 14 | 1724 | |||
AK301711.1 | - | 15 | 1633 | |||
AL833817.1 | - | 16 | 1868 | |||
BC016825.1 | - | 4 | 365 | |||
BC115005.1 | - | 16 | 1991 | 7782242 | ||
BC125206.1 | - | 15 | 1722 | 40123468 | ||
BC125207.1 | - | 14 | 1680 | 40123469 |