Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080431.2 | NP_001073900.1 | 4 | 1287 | ||
NM_001286646.1 | NP_001273575.1 | 4 | 1532 | |||
NM_001286648.1 | NP_001273577.1 | 4 | 1287 | |||
XM_011517169.2 | XP_011515471.1 | 5 | 1573 | |||
XM_011517170.2 | XP_011515472.1 | 6 | 2378 | |||
XM_011517171.2 | XP_011515473.1 | 5 | 1628 | |||
XM_011517172.2 | XP_011515474.1 | 5 | 2117 | |||
XM_011517173.2 | XP_011515475.1 | 5 | 1573 | |||
XM_011517174.2 | XP_011515476.1 | 4 | 1287 | |||
XM_011517176.2 | XP_011515478.1 | 4 | 1384 | |||
XM_011517177.2 | XP_011515479.1 | 4 | 1825 | |||
XM_011517178.2 | XP_011515480.1 | 4 | 4002 | |||
XM_011517179.2 | XP_011515481.1 | 4 | 1117 | |||
XM_017013648.1 | XP_016869137.1 | 5 | 2652 | |||
XM_017013649.1 | XP_016869138.1 | 5 | 1557 | |||
XM_017013650.1 | XP_016869139.1 | 4 | 2019 | |||
GenBank mRNA | AB032952.1 | - | 2 | 683 | ||
AK128212.1 | - | 4 | 1287 | |||
BC090891.1 | - | 4 | 1048 | |||
BC157840.1 | - | 4 | 1029 | 9057059 | ||
BC171836.1 | - | 4 | 1029 |