Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTUS1 SIRNA Assays ›
Gene Symbol:
MTUS1
Gene Name:
microtubule associated tumor suppressor 1
Gene Aliases:
ATBP, ATIP, ICIS, MP44, MTSG1
Chromosome Location:
Chr.8: 17643794 - 17801579 on Build GRCh38
UniGene ID:
Hs.7946
Species:
Human
siRNA ID
HSS148539
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTUS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001924.2
NP_001001924.1
9
3426
NM_001001925.2
NP_001001925.1
8
3264
NM_001001931.2
NP_001001931.1
6
1013
NM_001166393.1
NP_001159865.1
6
658
NM_020749.4
NP_065800.1
4
810
XM_005273577.3
XP_005273634.1
9
3338
XM_005273578.3
XP_005273635.1
9
3426
XM_005273579.3
XP_005273636.1
9
3338
XM_005273580.3
XP_005273637.1
8
3176
XM_005273581.3
XP_005273638.1
8
3264
XM_005273582.3
XP_005273639.1
8
3176
XM_011544595.2
XP_011542897.1
9
3338
XM_017013698.1
XP_016869187.1
10
3344
XM_017013699.1
XP_016869188.1
10
3566
XM_017013700.1
XP_016869189.1
9
3225
XM_017013701.1
XP_016869190.1
10
3342
XM_017013702.1
XP_016869191.1
8
2997
XM_017013704.1
XP_016869193.1
3
332
GenBank mRNA
AB033114.1
-
8
2878
AF121259.1
-
4
450
AF293357.1
-
4
723
AK024357.1
-
3
336
AK125188.1
-
6
1013
AK289750.1
-
4
809
AK294860.1
-
6
599
AK304105.1
-
7
895
AK314692.1
-
4
744
AL096842.1
-
9
2218
BC007328.2
-
8
912
BC117564.1
-
4
486
BC136320.1
-
9
3177
9020307
BC142971.1
-
4
743
BX648879.1
-
3
254
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-sq56x:80/10.239.161.141:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline