Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MTUS1 SIRNA Assays ›
Gene Symbol:
MTUS1
Gene Name:
microtubule associated tumor suppressor 1
Gene Aliases:
ATBP, ATIP, ICIS, MP44, MTSG1
Chromosome Location:
Chr.8: 17643794 - 17801579 on Build GRCh38
UniGene ID:
Hs.7946
Species:
Human
siRNA ID
HSS148541
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MTUS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001001924.2
NP_001001924.1
15
4250
NM_001001925.2
NP_001001925.1
14
4088
NM_001001931.2
NP_001001931.1
12
1837
NM_001166393.1
NP_001159865.1
12
1482
NM_020749.4
NP_065800.1
10
1634
XM_005273577.3
XP_005273634.1
15
4158
XM_005273578.3
XP_005273635.1
15
4246
XM_005273579.3
XP_005273636.1
15
4162
XM_005273580.3
XP_005273637.1
14
3996
XM_005273581.3
XP_005273638.1
14
4084
XM_005273582.3
XP_005273639.1
14
4000
XM_011544595.2
XP_011542897.1
15
4192
XM_017013698.1
XP_016869187.1
16
4164
XM_017013699.1
XP_016869188.1
16
4386
XM_017013700.1
XP_016869189.1
15
4049
XM_017013701.1
XP_016869190.1
16
4166
XM_017013702.1
XP_016869191.1
14
3821
XM_017013704.1
XP_016869193.1
9
1156
GenBank mRNA
AB033114.1
-
14
3702
AF086371.1
-
2
478
AF121259.1
-
10
1274
AF293357.1
-
10
1547
AK000172.1
-
3
375
AK024357.1
-
9
1160
AK093875.1
-
2
1993
AK125188.1
-
12
1837
AK289750.1
-
10
1633
AK294860.1
-
12
1423
AK304105.1
-
13
1719
AK314692.1
-
10
1568
AL096842.1
-
15
3042
BC033842.1
-
7
726
BC136320.1
-
15
4001
9020307
BC142971.1
-
10
1567
BX648879.1
-
8
2140
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0