Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM135A SIRNA Assays ›
Gene Symbol:
FAM135A
Gene Name:
family with sequence similarity 135 member A
Gene Aliases:
KIAA1411
Chromosome Location:
Chr.6: 70413404 - 70561174 on Build GRCh38
UniGene ID:
Hs.211700
Species:
Human
siRNA ID
HSS148574
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM135A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105531.2
NP_001099001.1
22
4334
NM_001162529.1
NP_001156001.1
20
4537
NM_020819.4
NP_065870.3
21
4197
XM_005248743.1
XP_005248800.1
23
4918
XM_005248744.4
XP_005248801.1
22
4885
XM_005248745.2
XP_005248802.1
22
4840
XM_005248748.2
XP_005248805.1
23
4330
XM_005248749.1
XP_005248806.1
24
4246
XM_005248751.2
XP_005248808.1
24
3658
XM_011535993.2
XP_011534295.1
22
4638
XM_011535994.2
XP_011534296.1
23
5058
XM_011535995.2
XP_011534297.1
23
4859
XM_011535996.2
XP_011534298.1
23
5016
XM_011535997.2
XP_011534299.1
21
4760
XM_017011124.1
XP_016866613.1
22
4891
XM_017011125.1
XP_016866614.1
21
4663
XM_017011126.1
XP_016866615.1
22
4977
XM_017011127.1
XP_016866616.1
21
4759
XM_017011128.1
XP_016866617.1
20
4683
XM_017011129.1
XP_016866618.1
23
4486
XM_017011130.1
XP_016866619.1
21
4133
XM_017011131.1
XP_016866620.1
22
4234
XM_017011132.1
XP_016866621.1
21
4221
XM_017011133.1
XP_016866622.1
20
4011
XM_017011134.1
XP_016866623.1
23
3580
XM_017011135.1
XP_016866624.1
21
4739
XM_017011136.1
XP_016866625.1
12
3466
GenBank mRNA
AB037832.1
-
20
4505
AJ420590.1
-
6
671
AK002067.1
-
4
436
AK128547.1
-
9
1900
AL136820.1
-
9
3162
BC065767.1
-
2
231
BC131782.1
-
22
4160
40124841
CR749254.1
-
21
4035
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cellular lipid metabolic process
carboxylic ester hydrolase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0