Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020950.1 | NP_066001.1 | 2 | 783 | ||
GenBank mRNA | AB046834.1 | - | 2 | 771 |