Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020956.2 | NP_066007.1 | 6 | 2805 | ||
NM_181882.2 | NP_870998.2 | 7 | 2156 | |||
XM_011527171.2 | XP_011525473.1 | 7 | 2148 | |||
XM_017027046.1 | XP_016882535.1 | 3 | 2307 | |||
XM_017027047.1 | XP_016882536.1 | 4 | 1850 | |||
GenBank mRNA | AB046840.1 | - | 3 | 2286 | ||
AF321191.1 | - | 7 | 2156 | |||
AF321192.1 | - | 6 | 2805 | |||
BC067266.1 | - | 7 | 2158 | 6177899 |