Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021623.1 | NP_067636.1 | 9 | 949 | ||
XM_011544605.2 | XP_011542907.1 | 9 | 834 | |||
XM_011544606.2 | XP_011542908.1 | 9 | 885 | |||
XM_011544607.2 | XP_011542909.1 | 9 | 847 | |||
XM_017013718.1 | XP_016869207.1 | 8 | 700 | |||
XM_017013719.1 | XP_016869208.1 | 8 | 670 | |||
XM_017013720.1 | XP_016869209.1 | 8 | 785 | |||
GenBank mRNA | AA985353.1 | - | 3 | 186 | ||
AF286164.1 | - | 9 | 874 | |||
AK291842.1 | - | 9 | 893 | |||
AK296068.1 | - | 8 | 742 | |||
AK309020.1 | - | 2 | 66 | |||
DC332022.1 | - | 2 | 66 |