Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022567.2 | NP_072089.1 | 2 | 1855 | ||
XM_017029709.1 | XP_016885198.1 | 3 | 1523 | |||
GenBank mRNA | AF254868.1 | - | 3 | 1523 | ||
AJ278865.1 | - | 2 | 1881 | |||
BC112242.1 | - | 2 | 1469 | 8327710 |