Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302643.1 | NP_001289572.1 | 5 | 701 | ||
NM_022082.3 | NP_071365.3 | 4 | 639 | |||
XM_011528978.2 | XP_011527280.1 | 4 | 551 | |||
GenBank mRNA | AK074107.1 | - | 4 | 575 | ||
AK095473.1 | - | 4 | 490 | |||
AK125597.1 | - | 5 | 644 | |||
AK298503.1 | - | 4 | 545 | |||
BC025312.1 | - | 5 | 641 | 4563089 | ||
BC038593.1 | - | 4 | 481 |