Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMRTA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032110.2 | NP_115486.1 | 3 | 1289 | ||
XM_011541937.2 | XP_011540239.1 | 2 | 2078 | |||
GenBank mRNA | AJ301580.1 | - | 2 | 783 | ||
BC136282.1 | - | 2 | 1045 | 9020269 | ||
BC143800.1 | - | 2 | 1045 | 9052314 |