Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022159.3 | NP_071442.2 | 2 | 272 | ||
GenBank mRNA | AB032017.1 | - | 2 | 233 | ||
AK292894.1 | - | 2 | 165 | |||
AK313738.1 | - | 2 | 271 | |||
AY358360.1 | - | 2 | 139 | |||
BC025721.1 | - | 2 | 138 | 5229055 | ||
DA477962.1 | - | 2 | 273 |
Set Membership: |
![]() |