Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MMP25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022468.4 | NP_071913.1 | 6 | 1102 | ||
XM_011522602.2 | XP_011520904.1 | 6 | 839 | |||
XM_011522604.2 | XP_011520906.1 | 5 | 953 | |||
XM_011522605.2 | XP_011520907.1 | 3 | 293 | |||
GenBank mRNA | AB042328.1 | - | 6 | 1034 | ||
AF145442.2 | - | 6 | 889 | |||
AF185270.1 | - | 6 | 889 | |||
AJ239053.1 | - | 6 | 1046 | |||
AJ272137.1 | - | 6 | 1102 |