Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCKSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023009.6 | NP_075385.1 | 2 | 778 | ||
NR_052852.1 | 2 | 553 | ||||
GenBank mRNA | AL713653.1 | - | 2 | 761 | ||
AY007131.1 | - | 2 | 761 | |||
BC007904.2 | - | 2 | 763 | 4138092 | ||
BC066915.1 | - | 2 | 772 | 5259353 | ||
DT218825.1 | - | 2 | 545 | |||
X70326.1 | - | 2 | 585 |