Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMUB2 SIRNA Assays ›
Gene Symbol:
TMUB2
Gene Name:
transmembrane and ubiquitin like domain containing 2
Gene Aliases:
FP2653
Chromosome Location:
Chr.17: 44186956 - 44191731 on Build GRCh38
UniGene ID:
Hs.181391
Species:
Human
siRNA ID
HSS149101
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMUB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001076674.1
NP_001070142.1
4
1030
NM_024107.2
NP_077012.2
3
962
NM_177441.2
NP_803190.2
3
1463
XM_011525198.2
XP_011523500.1
3
1366
XM_011525200.2
XP_011523502.1
5
1058
XM_011525201.2
XP_011523503.1
4
1038
XM_011525203.2
XP_011523505.1
5
1156
XM_011525204.2
XP_011523506.1
4
1095
XM_011525205.2
XP_011523507.1
4
1556
XM_011525206.2
XP_011523508.1
5
1242
XM_011525207.2
XP_011523509.1
4
1174
XM_011525208.2
XP_011523510.1
3
1067
XM_011525209.2
XP_011523511.1
3
1043
XM_011525210.2
XP_011523512.1
6
1132
XM_011525211.2
XP_011523513.1
4
889
XM_017025047.1
XP_016880536.1
4
1469
XM_017025048.1
XP_016880537.1
4
1139
XM_017025049.1
XP_016880538.1
3
1453
XM_017025050.1
XP_016880539.1
3
1067
XM_017025051.1
XP_016880540.1
3
985
XM_017025052.1
XP_016880541.1
5
1358
XM_017025053.1
XP_016880542.1
6
1045
XM_017025054.1
XP_016880543.1
4
1342
XM_017025055.1
XP_016880544.1
4
929
XM_017025056.1
XP_016880545.1
4
767
GenBank mRNA
AF289564.1
-
3
645
AF370373.1
-
3
1440
AK294198.1
-
6
1128
AL833899.1
-
3
1463
AY007092.1
-
3
885
AY358780.1
-
3
936
BC001792.1
-
3
687
BC004306.1
-
3
609
BC063489.1
-
3
936
5239969
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-k7c2b:80/10.239.160.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0