Gene Symbol: KREMEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253725.1 | NP_001240654.1 | 1, 2 | 410 | ||
NM_001253726.1 | NP_001240655.1 | 1, 2 | 410 | |||
NM_024507.3 | NP_078783.1 | 1, 2 | 410 | |||
NM_172229.2 | NP_757384.1 | 1, 2 | 410 | |||
GenBank mRNA | AB086355.1 | - | 1, 2 | 105 | ||
AB086356.1 | - | 1, 2 | 105 | |||
AB086357.1 | - | 1, 2 | 105 | |||
AB086405.1 | - | 1, 2 | 105 | |||
AK027669.1 | - | 1, 2 | 314 | |||
AK075033.1 | - | 1, 2 | 351 | |||
AK223422.1 | - | 1, 2 | 182 | |||
AK301953.1 | - | 1, 2 | 351 | |||
BC003533.1 | - | 1, 2 | 217 | 3609280 | ||
BC009383.2 | - | 1, 2 | 410 | 4127918 |