Gene Symbol: KREMEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253725.1 | NP_001240654.1 | 6 | 868 | ||
NM_001253726.1 | NP_001240655.1 | 6 | 868 | |||
NM_024507.3 | NP_078783.1 | 6 | 985 | |||
NM_172229.2 | NP_757384.1 | 6 | 985 | |||
GenBank mRNA | AB086355.1 | - | 6 | 680 | ||
AB086356.1 | - | 6 | 680 | |||
AB086357.1 | - | 6 | 680 | |||
AB086405.1 | - | 6 | 680 | |||
AK027669.1 | - | 6 | 889 | |||
AK075033.1 | - | 6 | 809 | |||
AK223422.1 | - | 6 | 757 | |||
AK301953.1 | - | 6 | 809 | |||
BC003533.1 | - | 6 | 792 | 3609280 | ||
BC009383.2 | - | 6 | 985 | 4127918 |