Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024516.3 | NP_078792.1 | 2 | 813 | ||
GenBank mRNA | BC003640.2 | - | 2 | 509 | 3356565 | |
EF195235.1 | - | 2 | 509 |