Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256541.1 | NP_001243470.1 | 2, 3 | 674 | ||
NM_024600.5 | NP_078876.2 | 1, 2 | 895 | |||
GenBank mRNA | AK024551.1 | - | 1, 2 | 896 | ||
AY358178.1 | - | 1, 2 | 322 | |||
AY676494.1 | - | 1, 2 | 896 | |||
BC004932.1 | - | 2, 3 | 674 | 3622114 | ||
BC105785.1 | - | 1, 2 | 395 | 4692469 |