Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PAQR6 SIRNA Assays ›
Gene Symbol:
PAQR6
Gene Name:
progestin and adipoQ receptor family member 6
Gene Aliases:
PRdelta
Chromosome Location:
Chr.1: 156243321 - 156248488 on Build GRCh38
UniGene ID:
Hs.235873
Species:
Human
siRNA ID
HSS149349
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PAQR6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001272104.1
NP_001259033.1
8
1063
NM_001272105.1
NP_001259034.1
9
1135
NM_001272106.1
NP_001259035.1
6
1029
NM_001272107.1
NP_001259036.1
7
1198
NM_001272108.1
NP_001259037.1
6
688
NM_001272109.1
NP_001259038.1
5
953
NM_001272110.1
NP_001259039.1
7
936
NM_001272111.1
NP_001259040.1
7
894
NM_001272112.1
NP_001259041.1
5
721
NM_001272113.1
NP_001259042.1
5
591
NM_024897.3
NP_079173.2
7
1269
NM_198406.2
NP_940798.1
8
1021
NR_073610.1
7
945
XM_005245494.3
XP_005245551.1
6
1189
XM_006711546.2
XP_006711609.1
8
1032
XM_006711547.3
XP_006711610.1
8
1505
XM_006711548.3
XP_006711611.1
7
1387
XM_006711552.2
XP_006711615.1
6
1130
XM_006711553.3
XP_006711616.1
6
1172
XM_011510000.2
XP_011508302.1
7
1295
XM_017002385.1
XP_016857874.1
7
2663
GenBank mRNA
AF455045.1
-
7
1117
AF455046.1
-
5
898
AF455047.1
-
7
860
AK026325.1
-
4
461
AK054682.1
-
7
875
AK094346.1
-
7
914
AK095143.1
-
8
993
AK095197.1
-
7
1204
AK294143.1
-
7
873
AK295915.1
-
7
887
AK295916.1
-
6
985
AK307441.1
-
6
725
AK316034.1
-
9
1074
AL834227.1
-
7
1089
AY424284.1
-
8
942
BC058509.1
-
8
905
5770270
BC071623.1
-
3
182
BI819919.1
-
5
642
BP362806.1
-
5
523
FJ200490.1
-
4
390
FJ200491.1
-
5
452
FJ200492.1
-
5
517
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
steroid hormone mediated signaling pathway
response to steroid hormone
steroid hormone receptor activity
steroid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0