Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002814.2 | NP_001002814.2 | 6 | 3875 | ||
NM_025151.4 | NP_079427.4 | 5 | 1973 | |||
GenBank mRNA | AF368294.1 | - | 5 | 1943 | ||
AK026177.1 | - | 2 | 214 | |||
AK092296.1 | - | 4 | 1281 | |||
AK122583.1 | - | 3 | 2014 | |||
AK124422.1 | - | 1 | 773 | |||
AK226111.1 | - | 6 | 3565 | |||
AK291781.1 | - | 5 | 1970 | |||
AL832155.1 | - | 3 | 1456 | |||
AL833169.1 | - | 3 | 1284 | |||
AY280966.1 | - | 3 | 2296 | |||
AY280968.1 | - | 6 | 3832 | |||
AY280969.1 | - | 2 | 3045 | |||
BC077720.1 | - | 5 | 1943 | 6537110 | ||
DQ083995.1 | - | 5 | 1697 | |||
DQ236342.1 | - | 5 | 1943 |