Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KCNIP4 SIRNA Assays ›
Gene Symbol:
KCNIP4
Gene Name:
potassium voltage-gated channel interacting protein 4
Gene Aliases:
CALP, KCHIP4
Chromosome Location:
Chr.4: 20728520 - 21948801 on Build GRCh38
UniGene ID:
Hs.655705
Species:
Human
siRNA ID
HSS149479
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KCNIP4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001035003.1
NP_001030175.1
5, 6
720
NM_001035004.1
NP_001030176.1
5, 6
641
NM_025221.5
NP_079497.2
6, 7
657
NM_147181.3
NP_671710.1
5, 6
555
NM_147182.3
NP_671711.1
6, 7
667
NM_147183.3
NP_671712.1
5, 6
474
XM_005248190.4
XP_005248247.1
5, 6
570
XM_011513882.2
XP_011512184.1
6, 7
701
XM_011513883.2
XP_011512185.1
5, 6
570
XM_011513884.2
XP_011512186.1
5, 6
594
XM_011513885.2
XP_011512187.1
5, 6
5890
XM_011513886.2
XP_011512188.1
5, 6
605
XM_011513887.2
XP_011512189.1
5, 6
1370
XM_011513889.2
XP_011512191.1
5, 6
560
XM_017008653.1
XP_016864142.1
5, 6
743
GenBank mRNA
AF302044.1
-
4, 5
435
AF305072.1
-
5, 6
537
AF367023.1
-
5, 6
587
AF367024.1
-
4, 5
485
AF453246.1
-
4, 5
351
AK289661.1
-
5, 6
549
AK289922.1
-
6, 7
627
AK290998.1
-
5, 6
557
AK294188.1
-
5, 6
570
AK314058.1
-
721
AY029176.1
-
5, 6
547
AY118170.1
-
5, 6
474
BC032520.1
-
5, 6
595
5494835
BI596848.1
-
661
BX648990.1
-
5, 6
721
DQ148487.1
-
5, 6
537
DQ148488.1
-
4, 5
435
DQ148489.1
-
5, 6
474
DQ148490.1
-
5, 6
720
DQ148491.1
-
5, 6
641
DQ148492.1
-
5, 6
486
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cardiac conduction
potassium ion transmembrane transport
protein localization to plasma membrane
regulation of potassium ion transmembrane transport
voltage-gated ion channel activity
potassium channel activity
calcium ion binding
potassium channel regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline